Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP C1 + C2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB098330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098330 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098330 Supplier Novus Biologicals Supplier No. NBP298634AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hnRNP C1 + C2 Polyclonal specifically detects hnRNP C1 + C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen hnRNP C1 + C2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias C1, C2, heterogeneous nuclear ribonucleoprotein C (C1/C2), heterogeneous nuclear ribonucleoproteins C1/C2, HNRNP, hnRNP C1/C2, hnRNPC, HNRPC, MGC104306, MGC105117, MGC117353, MGC131677, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein, SNRPC
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human hnRNP C1 + C2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 3183
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.