Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP F Antibody (OTI5F5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270907APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270907APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270907APC Supplier Novus Biologicals Supplier No. NBP270907APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP F Monoclonal specifically detects hnRNP F in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen hnRNP F
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5F5
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias heterogeneous nuclear ribonucleoprotein F, hnRNP F, HNRPFHnRNP F protein, mcs94-1, MGC110997, Nucleolin-like protein mcs94-1, OK/SW-cl.23
Gene Symbols HNRNPF
Host Species Mouse
Immunogen Full length recombinant protein of human hnRNP F (NP_004957) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 3185.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.