Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP G Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | hnRNP G |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179904
![]() |
Novus Biologicals
NBP179904 |
100 μL |
Each for $487.50
|
|
|||||
NBP17990420
![]() |
Novus Biologicals
NBP17990420UL |
20 μL | N/A | N/A | N/A | ||||
Description
hnRNP G Polyclonal specifically detects hnRNP G in Human samples. It is validated for Western Blot.Specifications
hnRNP G | |
Polyclonal | |
Rabbit | |
NP_002130 | |
27316 | |
The specific Immunogen is proprietary information. Peptide sequence SSSRDGYGGSRDSYSSSRSDLYSSGRDRVGRQERGLPPSMERGYPPPRDS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Glycoprotein p43, heterogeneous nuclear ribonucleoprotein G, hnRNP G, hnRNP-G, HNRPG, RBMXP1, RBMXRT, RNA binding motif protein, X chromosome, RNA binding motif protein, X-linked, RNA-binding motif protein, X chromosome, RNMX | |
RBMX | |
IgG | |
43 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title