Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP H Antibody, Novus Biologicals™
SDP

Catalog No. NB100385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100385 Supplier Novus Biologicals Supplier No. NB100385
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 15 publications

hnRNP H Polyclonal specifically detects hnRNP H in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen hnRNP H
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:30000, Simple Western 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 4-10 ug/mg of lysate, Immunohistochemistry-Paraffin 1:100
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_005511.1
Gene Alias DKFZp686A15170, heterogeneous nuclear ribonucleoprotein H, heterogeneous nuclear ribonucleoprotein H1 (H), hnRNP H, HNRPH, HNRPH1hnRNPH
Gene Symbols HNRNPH1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 400 and the C-terminus (residue 448) of human Heterogeneous Nuclear Ribonucleoprotein H1 using the numbering given in entry NP_005511.1 (GeneID 3187).
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 3187
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Gray short-tailed opossum.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.