Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP K Antibody (N10 P2D3*G2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20424A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20424A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20424A32 Supplier Novus Biologicals Supplier No. NBP250424AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP K Monoclonal specifically detects hnRNP K in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen hnRNP K
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone N10 P2D3*G2
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP
Gene Symbols HNRNPK
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide SVKQYSGKFF (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3190
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.