Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP K Antibody (N10 P2D3*G2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309422 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309422 Supplier Novus Biologicals Supplier No. NBP250424MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP K Monoclonal antibody specifically detects hnRNP K in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen hnRNP K
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone N10 P2D3*G2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide SVKQYSGKFF (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3190
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.