Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP L Antibody (4D11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB1206106G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206106G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206106G Supplier Novus Biologicals Supplier No. NB1206106G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen hnRNP L
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4D11
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Accession No. P14866
Gene Alias FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL
Gene Symbols HNRNPL
Host Species Mouse
Immunogen Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3191
Test Specificity Human, Mouse, Rat, Bovine, Monkey, Chicken and Newt.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.