Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP-R Antibody, Novus Biologicals™
SDP

Catalog No. NBP15715820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15715820 20 μL
NBP157158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15715820 Supplier Novus Biologicals Supplier No. NBP15715820UL

Rabbit Polyclonal Antibody

hnRNP-R Polyclonal specifically detects hnRNP-R in Human samples. It is validated for Western Blot.

Specifications

Antigen hnRNP-R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. O43390
Gene Alias FLJ25714, heterogeneous nuclear ribonucleoprotein R, hnRNP R, HNRPRhnRNP-R
Gene Symbols HNRNPR
Host Species Rabbit
Immunogen Synthetic peptides corresponding to HNRNPR(heterogeneous nuclear ribonucleoprotein R) The peptide sequence was selected from the N terminal of HNRNPR. Peptide sequence ANQVNGNAVQLKEEEEPMDTSSVTHTEHYKTLIEAGLPQKVAERLDEIFQ.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10236
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.