Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP-R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | hnRNP-R |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157158
![]() |
Novus Biologicals
NBP157158 |
100 μL |
Each for $487.50
|
|
|||||
NBP15715820
![]() |
Novus Biologicals
NBP15715820UL |
20 μL | N/A | N/A | N/A | ||||
Description
hnRNP-R Polyclonal specifically detects hnRNP-R in Human samples. It is validated for Western Blot.Specifications
hnRNP-R | |
Polyclonal | |
Rabbit | |
O43390 | |
10236 | |
Synthetic peptides corresponding to HNRNPR(heterogeneous nuclear ribonucleoprotein R) The peptide sequence was selected from the N terminal of HNRNPR. Peptide sequence ANQVNGNAVQLKEEEEPMDTSSVTHTEHYKTLIEAGLPQKVAERLDEIFQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ25714, heterogeneous nuclear ribonucleoprotein R, hnRNP R, HNRPRhnRNP-R | |
HNRNPR | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title