Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP U Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292501 Supplier Novus Biologicals Supplier No. NB1002135PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hnRNP U Polyclonal specifically detects hnRNP U in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen hnRNP U
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRPU, p120, p120 nuclear protein, pp120, SAFA, SAF-AhnRNPU, Scaffold attachment factor A, U21.1
Host Species Rabbit
Immunogen Maps to a region between residue 775 and the C-terminus (residue 824) of human Heterogeneous Nuclear Ribonucleoprotein U using the numbering given in entry NP_114032.1 (GeneID 3192).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3192
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.