Learn More
hnRNP UL1 Monoclonal Antibody (EAP 4A11), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$1592.00
Specifications
| Antigen | hnRNP UL1 |
|---|---|
| Clone | EAP 4A11 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-980-637
|
Invitrogen™
14980637 |
2 mg |
Each of 1 for $1,592.00
|
|
|||||
Description
Description: This EAP 4A11 monoclonal antibody recognizes human hnRNP UL1. EAP 4A11 detects a ∽95 kDa band corresponding to hnRNP UL1 by Western Blot and stains hnRNP UL1 in the nucleus in Immunocytochemistry. Applications Reported: This EAP 4A11 antibody has been reported for use in western blot and immunocytochemistry/immunofluorescence. Applications Tested: This EAP 4A11 antibody has been tested by western blot of human cell lysates and immunocytochemistry/immunofluorescence of human cell lines. This may be used at less than or equal to 4.0 μg/ml for western blot and less than or equal to 2.5 μg/mL for immunocytochemistry/immunofluorescence.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.Specifications
| hnRNP UL1 | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rat | |
| Human | |
| Q9BUJ2 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| EAP 4A11 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 11100 | |
| Primary | |
| 4°C, do not freeze | |
| Hnrnpul1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.