Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 50555224 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-555-224 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-555-224 Supplier Proteintech Group Inc Supplier No. 127701AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hnRNP D/AUF1 binds with high affinity to RNA molecules that contain AU-rich elements (AREs) found within the 3′ untranslated regions of many protooncogenes and cytokine mRNAs. It also binds to double and single stranded DNA sequences in a specific manner and functions a transcription factor. Each of the RNA-binding domains specifically can bind solely to a single stranded nonmonotonous 5′-UUAG-3′ sequence and also weaker to the single-stranded 5′-TTAGGG-3′ telomeric DNA repeat. It may be involved in translationally coupled mRNA turnover (referenced from swissprot).

Specifications

Antigen HNRNPD
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HNRNPD
Gene Accession No. Q14103, Q60668, Q9JJ54
Gene Alias AUF1, AUF1A, hnRNP D0, HNRNPD, hnRNPD0, HNRPD, P37
Gene Symbols HNRNPD
Host Species Rabbit
Immunogen HNRNPD Fusion Protein Ag3458
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11991, 3184, 79256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.