Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPUL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237687 Supplier Novus Biologicals Supplier No. NBP237687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HNRNPUL2 Polyclonal specifically detects HNRNPUL2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen HNRNPUL2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_001073027.1
Gene Alias DKFZp762N1910, heterogeneous nuclear ribonucleoprotein U-like 2, heterogeneous nuclear ribonucleoprotein U-like protein 2, HNRPUL2, SAF-A2, Scaffold-attachment factor A2
Gene Symbols HNRNPUL2
Host Species Rabbit
Immunogen This antibody maps to a region between residue 25 to 75 of human Heterogeneous Nuclear Ribonucleoprotein U-Like 2 using the numbering given in entry NP_001073027.1 (GeneID 221092).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221092
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.