Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-1/HMOX1/HSP32 Antibody (6B8-2F2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP259338G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259338G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259338G Supplier Novus Biologicals Supplier No. NBP259338G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HO-1/HMOX1/HSP32
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6B8-2F2
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Gene Symbols HMOX1
Host Species Mouse
Immunogen His-tagged Rat HO-1
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3162
Test Specificity Detects 32kDa. Does not cross-react with HO-2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.