Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-1/HMOX1/HSP32 Antibody (HO-1-1), Novus Biologicals™
SDP

Catalog No. NBP197507SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197507SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197507SS Supplier Novus Biologicals Supplier No. NBP197507SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 14 publications

HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HO-1/HMOX1/HSP32
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HO-1-1
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western, Flow Cytometry 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P09601
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Gene Symbols HMOX1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1.
Molecular Weight of Antigen 33 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3162
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.