Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-1/HMOX1/HSP32 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB338040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338040 Supplier Novus Biologicals Supplier No. NBP177459PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HO-1/HMOX1/HSP32 Polyclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen HO-1/HMOX1/HSP32
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Heme Oxygenase 1 protein (between residues 100-150) [UniProt# P09601]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3162
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.