Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HO-1/HMOX1/HSP32 Native Protein
SDP

Catalog No. p-7249119 Shop All R&D Systems Products
Click to view available options
Quantity:
100 μg
200 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Native Protein

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 20mM Tris pH7.5, 0.1mM EDTA, 0.1% triton x-100, 0.2% Na cholate, <50mM KCl
Gene ID (Entrez) 3162
Molecular Weight (g/mol) M.W. Theoretical: 33 kDa
Quantity 50 μg
Storage Requirements Store at -70C. Avoid freeze-thaw cycles.
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Gene Symbol HMOX1
Research Category Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Purity or Quality Grade >85%
Protein HO-1/HMOX1/HSP32
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.