Learn More
Description
Hoechst Janelia Fluor™ 646 is a desirable alternative to large oligonucleotide-conjugated antibodies for PAINT experiments, particularly for bacterial studies. Hoechst Janelia Fluor™ 646 exhibits a quantum yield (QY) of 0.54; extinction coefficicent 152, 000; excitation maximum = 655 nm; emission maximum = 670 nm.
Fluorogenic: fluoresces only once bound to DNA, enabling hassle-free no-wash experiments. Hoechst Janelia Fluor™ 646 is suitable for multicolor microscopy experiments and for use in live cell imaging.
To measure the absorbance spectrum of Hoechst Janelia Fluo™ 646 we recommend the following solvent: TFE plus 0.1% TFA.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2-(2-(4-(4-(5-(4-methylpiperazin-1-yl)-1 H,3 'H-[2,5 '-bibenzo[d]imidazol]-2 '-yl)phenoxy)butanamido)ethoxy)ethoxy)ethyl)carbamoyl)benzoat |
| CAS | 2462011-60-3 |
| Quantity | 100 μg |
| Product Line | Tocris |
| Target | Fluorescent DNA Stains |
| Molecular Formula | C64H70N10O7Si |
| Purity | 0.90 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

