Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
SDP

Catalog No. 03500174
Encompass
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable

Specifications

For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 1 Prep, 1 Reference
Thickness (English) 0.02 in.
Thickness (Metric) 0.07 cm
Volume (Metric) Well 51μL/ mm
Width (English) Well 2.6 in.
Width (Metric) Well 6.8 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.