Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
Specifications
| For Use With (Equipment) | SE 250, SE 260, MiniVE™ Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 10 |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.1 cm |
| Volume (Metric) Well | 4.8μL/ mm |
| Width (Metric) | 0.48 cm |
| Depth (English) | 0.51 in. |
| Depth (Metric) | 1.3 cm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction