Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
SDP

Catalog No. 03500192
Encompass
Click to view available options
No. of Wells:
1 Prep, 1 Reference
1 Prep, 2 Reference
12
15
20
28
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
11.5μL/ mm
120μL/ mm
171μL/ mm
3.1μL/ mm
4.1μL/ mm
4.3μL/ mm
5.7μL/ mm
6.2μL/ mm
7.7μL/ mm
Type:
Comb
Standard Comb

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth

Specifications

For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 20
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Volume (Metric) Well 4.1μL/ mm
Width (Metric) 0.41 cm
Depth (English) 0.98 in.
Depth (Metric) 2.5 cm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.