Hoefer™ HE 33 Mini Submarine Electrophoresis Unit Available on GSA/VA Contract for Federal Government customers only.

Designed for very fast separations of DNA restriction fragments in agarose gels

Manufacturer: Hoefer™ HE33B

 View more versions of this product

Catalog No. 03-500-131

  Check Availability
Add to Cart

    Specifications

    Temperature (Metric) Maximum 50°C
    Height (Metric) Exterior 7cm
    Length (English) Exterior 9.4 in.
    Length (Metric) Exterior 24cm
    Width (English) Exterior 5.1 in.
    Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
    Volume (English) Buffer 8.4 oz.
    Volume (Metric) Buffer 250mL
    Length (English) Gel 3.9 in.
    Width (Metric) Gel 7cm
    Amperage 500mA
    Dimensions (L x W x H) Exterior 9.4 x 5.1 x 2.8 in. (24 x 13 x 7cm)
    Humidity Range Up to 80%
    Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level
    Sample Size Up to 32
    Temperature (English) Maximum 122°F
    Voltage 500V
    Wattage 15w
    Product Type Mini horizontal Agarose electrophoresis unit
    Width (Metric) Exterior 13cm
    Length (Metric) Gel 10cm
    Width (English) Gel 2.7 in.
    Height (English) Exterior 2.8 in.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit