Learn More
Hoefer™ HE-PLUS Casting Kits
Click to view available options
Type:
Mini Casting Kit
Standard Casting Kit
Description
Includes
Mini Casting Kit includes four gel trays (6 × 6cm), two combs (5/8 wells) and casting stand. Standard Casting Kit includes one large gel tray (12.5 × 13cm), two medium gel trays (12.5 × 6cm), four combs (14/28 wells) and casting stand.
- Accessories and Replacement Parts for HE Systems
Specifications
Specifications
For Use With (Equipment) | HE-plus Horizontal Electrophoresis System |
Type | Standard Casting Kit |
Gel Dimensions | 12.5 x 13 cm |
Includes | One large gel tray (12.5 x 13cm), two medium gel trays (12.5 x 6cm), four combs (14/28-well) and casting stand |