Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ PVDF Membrane
SDP

Catalog No. 03500607
Encompass
Click to view available options
Length (Metric):
30 cm

Ideal for use in protein binding applications such as Western blots, solid phase assays, and immunoblotting procedures

  • Superior strength—withstands aggressive handling or can be used with automated equipment without breaking or tearing
  • Membrane binds a wide range of fragment sizes
  • Excellent sensitivity to detect low-level components
  • Low extractables
  • Hydrophobic
  • Stringent quality tests ensure lot-to-lot consistency and dependable results

Specifications

Length (Metric) 30 cm
Width (Metric) 3 m
Format Roll
Material PVDF
Pore Size 0.45 μm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.