Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hoefer™ SE 600 Series Complete Vertical Electrophoresis System


Click to view available options
Voltage:
1000 V
Description
- Offers durable construction for proven decades of performance
- Run one gel or two gels simultaneously, four with optional divider plates
- Produce straight lanes and sharp, well-defined bands—no “smiling”
- Provide two mechanisms to dissipate heat
- Gel sandwiches are completely immersed in the lower buffer so that heat is transferred evenly on both sides of the gel
- A heat exchanger suspended vertically in the lower chamber cools or heats buffer evenly
Suitable for denaturing and native polyacrylamide, 2-D, isoelectric focusing and agarose gel electrophoresis
Certifications
UL3101-1, CSA C22.2 1010.1, CE Certified, EN61010-1
Specifications
Specifications
Certifications/Compliance | EN61010-1, UL3101-1, CSA C22.2 1010.1, CE |
Humidity Range | Up to 80% |
Includes | Lower Buffer chamber, upper Buffer chamber, safety lid with high voltage Leads, heat exchanger, Glass plates, clamp assemblies, cams, slotted gaskets for upper Buffer chamber, dual gel casting stand with leveling base, bubble level, laminated gaskets for |
Temperature (English) Maximum | 113°F |
Temperature (Metric) Maximum | 45°C |
Temperature (Metric) Storage | 15°C to 30°C |
Thickness (English) Spacer | 0.05 in. |
Thickness (Metric) Spacer | 0.15 cm |
Volume (English) Buffer | 169.07 oz. |
Volume (Metric) Buffer | 5000 mL |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction