Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ TE42 and TE62 Tank Blotting Units
SDP

Catalog No. p-3004003
Encompass
Click to view available options
Type:
Blotting Unit
Includes:
4 gel cassettes, 12 total sponges

Perform high-intensity protein transfers in one hour

  • Over 1.5 m of platinum wire per electrode provides uniform field strength for efficient transfers
  • Hinged, color-coded cassettes enable simple loading and easy orientation of gel sandwich
  • Buffer volumes of 4 to 5 L required, depending on number of cassettes processed
  • Model TE62 offers a built-in heat exchanger in the base of the unit
  • Model TE42 can be used with optional heat exchanger (Mfr. No. TE47)

Certifications

CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1

Specifications

Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Content And Storage Lower chamber with built-in heat exchanger, safety lid with high voltage Leads, 2 electrode panels, four gel cassettes, eight 0.13 in. thick Foam sponges, four 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook
Device Holds Up to 4, 15 x 21 cm Gel or 16, 7 x 10 cm Gel
Type Blotting Unit
Humidity Range Up to 80%
Includes 4 gel cassettes, 12 total sponges
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (English) Buffer 135.25 to 169.07 oz.
Volume (Metric) Buffer 4000 to 5000 mL
Dimensions (L x W x H) 12.59 x 11.02 x 6.49 in. (32 x 28 x 16.5 cm)
Length (English) 12.59 in.
Length (Metric) 32 cm
Width (English) 11.02 in.
Width (Metric) 28 cm
Height (English) 6.49 in.
Height (Metric) 16.5 cm
Voltage 100 V
Wattage 200 w
Amperage 2 A
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.