Learn More
Eisco™ Hoffman Electrolysis Apparatus with PTFE Stopcock
SCI_ED
Catalog No. S41739 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S41739 1 Ea.
1 options
Catalog No. S41739 Supplier Eisco™ Supplier No. CH0740B
Add to Cart
Add to Cart
Hoffman Electrolysis Apparatus with PTFE Stopcock provides an effective and quantifiable method for electrolysis of water into hydrogen and oxygen.
  • Made of borosilicate 3.3 grade glass.
  • Graduated
  • Includes:
    • (2) Carbon electrodes in rubber stoppers
    • (2) Platinum electrodes in rubber stoppers
  • Ring stand and clamp required, not included (S41741 recomended)
  • Instructions included
Warranty and Services
1 year limited manufacturer warranty
Specifications
Type Electrolysis Apparatus
Quantity 1 Ea.
Material Borosilicate Glass
Includes Glass hoffman electrolysis apparatus with PTFE stopcocks, 2 platinum electrodes in rubber stoppers, 2 carbon electrodes in rubber stoppers
Length (Metric) 29 cm
For Use With (Application) Electroysis
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov