Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOIP/RNF31 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227290SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227290SS 0.025 mg
NBP227290 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227290SS Supplier Novus Biologicals Supplier No. NBP227290SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOIP/RNF31 Polyclonal specifically detects HOIP/RNF31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HOIP/RNF31
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Symbols RNF31
Host Species Rabbit
Immunogen A portion of amino acids 950-1000 from human RNF31 (HOIP) was used as the immunogen for this antibody. NP-060469.4
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55072
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in new world monkey, horse, naked mole rat, crab-eating macaque, rhesus monkey, white-cheeked gibbon, common chimpanzee, and sumatran orangutan, 88% in guinea pig, dog, common rabbit, and pig, 82% in panda, Chinese hamster, african bush elephant, gray short-tailed opossum, platypus, rat, and clawed frog.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.