Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOIP/RNF31 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen HOIP/RNF31
Concentration 0.5 mg/mL
Dilution Western Blot, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227290SS
SDP
View Documents
Novus Biologicals
NBP227290SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227290
SDP
View Documents
Novus Biologicals
NBP227290
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

HOIP/RNF31 Polyclonal specifically detects HOIP/RNF31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HOIP/RNF31
Western Blot, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
55072
A portion of amino acids 950-1000 from human RNF31 (HOIP) was used as the immunogen for this antibody. NP-060469.4
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein
RNF31
IgG
Protein A purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in new world monkey, horse, naked mole rat, crab-eating macaque, rhesus monkey, white-cheeked gibbon, common chimpanzee, and sumatran orangutan, 88% in guinea pig, dog, common rabbit, and pig, 82% in panda, Chinese hamster, african bush elephant, gray short-tailed opossum, platypus, rat, and clawed frog.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.