Learn More
Eisco™ Hollow Glass Prisms
SCI_ED
Catalog No. p-8677005 Shop All Eisco Products
Click to view available options
Product Type:
Hollow Cube
Hollow Prism Cube
Hollow Pyramidal
Hollow Rectangular, Slab
Length (Metric):
2.5 cm
8.29 cm
8.9 cm
10.2 cm
Catalog No. S27411
Supplier: Eisco™ PH0552I
Only null left
Add to Cart
Add to Cart
Hollow glass prisms are made from glass plates that are cemented together. Stoppers included. Excellent tools for optical experimentation as well as for studying Snells Law of Refraction. Change the liquids or gases in the prisms to change the refraction properties
  • Made from glass plates, cemented together
  • Stopper included
  • Hollow inside allows for a variety of optic experiments
  • Excellent tool for studying Snells Law of Refraction
  • Designed for educational use
Specifications
Product Type Hollow Prism Cube
Material Acrylic
For Use With (Application) Optics Experiments, Snell's Law of Refraction
Includes Prism, Stopper
Quantity 1 Ea.
Length (Metric) 2.5 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.