Learn More
Homeotic protein antennapedia Rabbit anti-Drosophila, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Homeotic protein antennapedia |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | ANTP |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide corresponding to a region of Fruit fly (NP_996176). Peptide sequence QQQQQPVVYASCKLQAAVGGLGMVPEGGSPPLVDQMSGHHMNAQMTLPHH |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.