Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER1 Antibody (2F7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2060A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2060A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2060A30 Supplier Novus Biologicals Supplier No. NBP250605AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HOMER1 Monoclonal specifically detects HOMER1 in Drosophila samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HOMER1
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2F7
Conjugate Alexa Fluor 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1
Gene Symbols HOMER1
Host Species Mouse
Immunogen GST-Drosophila HOMER1 (full-length) fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9456
Target Species Drosophila
Content And Storage Store at 4°C in the dark.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.