Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER2 Antibody (OTI1H3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271465A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271465A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271465A700 Supplier Novus Biologicals Supplier No. NBP271465AF700

Mouse Monoclonal Antibody

HOMER2 Monoclonal specifically detects HOMER2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HOMER2
Applications Western Blot
Classification Monoclonal
Clone OTI1H3
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACPD, CPD, cupidin, homer homolog 2 (Drosophila), homer homolog 3, homer protein homolog 2, homer, neuronal immediate early gene, 2, Homer-2, HOMER2A, HOMER-2A, HOMER2B, HOMER-2B, Vesl-2
Gene Symbols HOMER2
Host Species Mouse
Immunogen Full length recombinant protein of human HOMER2 (NP_004830) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9455.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.