Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Homoharringtonine
GREENER_CHOICESDP

Catalog No. 141610 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-161-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-161-0 Supplier Tocris Bioscience™ Supplier No. 1416/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of protein synthesis; antileukemic agent

Inhibitor of protein synthesis. Blocks elongation phase of translation by binding to the 60-S ribosome subunit. Antileukemic.

Chemical Identifiers

CAS 26833-87-4
Molecular Formula C29H39NO9

Specifications

Quantity 10 mg
Formula Weight 545.63
Purity >99%
Molecular Weight (g/mol) 545.63
Chemical Name or Material Homoharringtonine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.