Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Analytics™ GasAlertMax XT II Multigas Detectors
Monitor up to four hazardous gases with a robust, motorized pump for intelligent, remote sampling. The GasAlertMax XT II Multigas Detector is compact, comfortable, field tough, and ideal for confined sampling.
Supplier: Honeywell Analytics™ XTXW0MYNA
Description
- Integrated sampling pump with reliable diaphragm technology
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Equipped with standard datalogging and event logging
- Continuous LCD shows real-time gas concentrations
- Block detection with semi-conductor pressure sensor
- User adjustable alarm setpoints
- Multi-language support
- Water-resistant
- Size: 13.1 x 7.0 x 5.2cm (5.1 x 2.8 x 2.0 in.)
- Weight: 316g (11.1 oz.)
- Temperature Range: -20° to +50°C (-4° to +122°F)
- Humidity: 10 to 100% RH (non-condensing)
H2S Sensors
- Measuring Range: 0 to 200ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 1000ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 30% by vol
- Resolution: 0.1%
- Measuring Range: 0 to 100% LEL; 0 to 5.0% v/v
- Resolution: 1.0% LEL; 0.1% v/v
Warranty and Services
Full 2 year warranty including all sensors

Specifications
Audible, visual, vibrating | |
Yellow | |
5.08 cm | |
LCD | |
13 cm | |
O2: 0.1%, LEL: 1%, CO: 1ppm | |
-20°C to +50°C | |
11.1 oz. | |
2.8 in. |
8 to 13 hr. | |
2 in. | |
Carbon Monoxide, LEL, Oxygen | |
5.12 in. | |
10 to 100% | |
-4°F to +122°F | |
Full 2 year warranty including all sensors | |
316g | |
7.2 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction