Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlertMax XT II Multigas Detectors

Monitor up to four hazardous gases with a robust, motorized pump for intelligent, remote sampling. The GasAlertMax XT II Multigas Detector is compact, comfortable, field tough, and ideal for confined sampling.

Supplier:  Honeywell Analytics™ XTXWH0BNA

 View more versions of this product

Catalog No. 19-413-387


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

  • Integrated sampling pump with reliable diaphragm technology
  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
  • Equipped with standard datalogging and event logging
  • Continuous LCD shows real-time gas concentrations
  • Block detection with semi-conductor pressure sensor
  • User adjustable alarm setpoints
  • Multi-language support
  • Water-resistant
  • Size: 13.1 x 7.0 x 5.2cm (5.1 x 2.8 x 2.0 in.)
  • Weight: 316g (11.1 oz.)
  • Temperature Range: -20° to +50°C (-4° to +122°F)
  • Humidity: 10 to 100% RH (non-condensing)

 

H2S Sensors

  • Measuring Range: 0 to 200ppm
  • Resolution: 1.0ppm
CO Sensors
  • Measuring Range: 0 to 1000ppm
  • Resolution: 1.0ppm
O2 Sensors
  • Measuring Range: 0 to 30% by vol
  • Resolution: 0.1%
Combustible Gas Sensors
  • Measuring Range: 0 to 100% LEL; 0 to 5.0% v/v
  • Resolution: 1.0% LEL; 0.1% v/v

Warranty and Services

Full 2 year warranty including all sensors

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Audible, visual, vibrating
Black
5.08 cm
LCD
13 cm
O2: 0.1%, LEL: 1%, H2S: 1ppm
-20°C to +50°C
11.1 oz.
2.8 in.
8 to 13 hr.
2 in.
Hydrogen Sulfide, LEL, Oxygen
5.12 in.
10 to 100%
-4°F to +122°F
Full 2 year warranty including all sensors
316g
7.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.