Learn More
Honeywell Analytics™ GasAlertMax XT II Multigas Detectors
Monitor up to four hazardous gases with a robust, motorized pump for intelligent, remote sampling. The GasAlertMax XT II Multigas Detector is compact, comfortable, field tough, and ideal for confined sampling.
Supplier: Honeywell Analytics™ XTX00MBNA
Description
- Integrated sampling pump with reliable diaphragm technology
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Equipped with standard datalogging and event logging
- Continuous LCD shows real-time gas concentrations
- Block detection with semi-conductor pressure sensor
- User adjustable alarm setpoints
- Multi-language support
- Water-resistant
- Size: 13.1 x 7.0 x 5.2cm (5.1 x 2.8 x 2.0 in.)
- Weight: 316g (11.1 oz.)
- Temperature Range: -20° to +50°C (-4° to +122°F)
- Humidity: 10 to 100% RH (non-condensing)
H2S Sensors
- Measuring Range: 0 to 200ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 1000ppm
- Resolution: 1.0ppm
- Measuring Range: 0 to 30% by vol
- Resolution: 0.1%
- Measuring Range: 0 to 100% LEL; 0 to 5.0% v/v
- Resolution: 1.0% LEL; 0.1% v/v
Warranty and Services
Full 2 year warranty including all sensors

Specifications
Audible, visual, vibrating | |
Black | |
5.08 cm | |
LCD | |
13 cm | |
O2: 0.1%, CO: 1ppm | |
-20°C to +50°C | |
11.1 oz. | |
2.8 in. |
8 to 13 hr. | |
2 in. | |
Carbon Monoxide, Oxygen | |
5.12 in. | |
10 to 100% | |
-4°F to +122°F | |
Full 2 year warranty including all sensors | |
316g | |
7.2 cm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.