Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOOK3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017301 Supplier Novus Biologicals Supplier No. NBP281990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOOK3 Polyclonal specifically detects HOOK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HOOK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ31058, hHK3, HK3h-hook3, hook homolog 3 (Drosophila), protein Hook homolog 3
Gene Symbols HOOK3
Host Species Rabbit
Immunogen HOOK3 antibody was raised against a 19 amino acid peptide from near the amino terminus of human HOOK3. The immunogen is located within amino acids 210 - 260 of HOOK3. Amino Acid Squence: MERLNQSDSIEDPNSPAGR
Molecular Weight of Antigen 77 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84376
Test Specificity HOOK3 antibody is human, mouse and rat reactive. HOOK3 antibody is predicted to not cross-react with HOOK1 and HOOK2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.