Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOP Antibody (DS14F5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

Supplier:  Novus Biologicals NBP1975030.2MG

 View more versions of this product

Catalog No. NBP19750302


Only null left
Add to Cart

Description

Description

HOP Monoclonal specifically detects HOP in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Mustelid, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

HOP
Monoclonal
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
HOPX
Chicken HOP (p60).
0.2 mg
Primary
Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Hamster, Mustelid, Primate, Rabbit, Sheep, Xenopus species
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
DS14F5
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO
Mouse
Protein G purified
RUO
84525
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only