Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOP Antibody (DS14F5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$269.00 - $743.50

Specifications

Antigen HOP
Clone DS14F5
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018793
SDP
View Documents
Novus Biologicals
NBP1975030.025MG
0.025 mg
Each for $269.00
Only null left
Add to Cart
 
NBP19750305
SDP
View Documents
Novus Biologicals
NBP1975030.05MG
0.05 mg
Each for $390.00
Only null left
Add to Cart
 
NBP19750302
SDP
View Documents
Novus Biologicals
NBP1975030.2MG
0.2 mg
Each for $743.50
Only null left
Add to Cart
 
Description

Description

HOP Monoclonal specifically detects HOP in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Mustelid, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

HOP
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
84525
Chicken HOP (p60).
Primary
Store at -20C. Avoid freeze-thaw cycles.
DS14F5
Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Rat, Xenopus species, Canine, Chicken, Hamster, Rabbit, Pig, Mustelid, Primate, Mouse, Sheep, Bovine, Guinea Pig
CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO
HOPX
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.