Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific Horizontal Standard Midi-Gel Kits
Shop All CBS Scientific Products

Click to view available options
Gel Dimensions:
10 x 14 cm
11 x 10.5 cm
16 x 14 cm
Color:
Green
Pink
Yellow
Description
CBS Midi Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
- Platinum electrode cassettes can be easily removed for cleaning or replacement
- High quality, heavy duty construction with vast array of accessory options
- Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
- Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
- MHU-602 Midi-Horizontal runs up to a full microtitre plate (108 samples) per gel
- Tapered baffles allow easy in situ gel formation with or without gel tray
- Run up to 32 samples per gel. Two slots in each gel tray
- Combs are printed with catalog number and thickness for easy identification
- Choice of double-sided adjustable height combs gives researcher more options
- Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
- Horizontal Midi-Gels are available in four vivid colors; green, yellow, blue or fluorescent pink
- Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Specifications
Specifications
| Certifications/Compliance | CE |
| Color | Green |
| Description | Compatible Gel Tray: WGT-600 |
| Type | Midi-Gel Kit |
| Gel Dimensions | 16 x 14 cm |
| Volume (Metric) Buffer | 750 mL |
| Warranty | 3 Years |
| Dimensions (L x W) | Gel Tray I.D.: 16 x 12.7 cm (6.2 x 5 in.) |
| Dimensions (L x W x H) | 27.5 x 16.7 x 14 cm (10.8 x 6.5 x 5.5 in.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction