Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific Horizontal Standard Midi-Gel Kits
SCI_ED

Catalog No. p-6629092 Shop All CBS Scientific Products
Click to view available options
Gel Dimensions:
10 x 14 cm
11 x 10.5 cm
16 x 14 cm
Color:
Green
Pink
Yellow

Used with Midi Horizontal System.

CBS Midi Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
  • Platinum electrode cassettes can be easily removed for cleaning or replacement
  • High quality, heavy duty construction with vast array of accessory options
  • Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
  • Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
  • MHU-602 Midi-Horizontal runs up to a full microtitre plate (108 samples) per gel
  • Tapered baffles allow easy in situ gel formation with or without gel tray
  • Run up to 32 samples per gel. Two slots in each gel tray
  • Combs are printed with catalog number and thickness for easy identification
  • Choice of double-sided adjustable height combs gives researcher more options
  • Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
  • Horizontal Midi-Gels are available in four vivid colors; green, yellow, blue or fluorescent pink
  • Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Applications: Seperation and identification of DNA fragments, RNA fragments, PCR Products and synthetic oligonucleotides

Specifications

Certifications/Compliance CE
Color Yellow
Description Compatible Gel Tray: WGT-500
Type Midi-Gel Kit
Gel Dimensions 10 x 14 cm
Volume (Metric) Buffer 700 mL
Warranty 3 Years
Dimensions (L x W) Gel Tray I.D.: 10 x 12.7 cm (3.9 x 5 in.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.