Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORMAD2 Antibody (OTI3E4), PE, Novus Biologicals™
SDP

Catalog No. NB288165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288165 Supplier Novus Biologicals Supplier No. NBP272416PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HORMAD2 Monoclonal antibody specifically detects HORMAD2 in Human samples. It is validated for Western Blot

Specifications

Antigen HORMAD2
Applications Western Blot
Classification Monoclonal
Clone OTI3E4
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias HORMA domain containing 2, HORMA domain-containing protein 2, MGC26710
Host Species Mouse
Immunogen Full length human recombinant protein of human HORMAD2(NP_689723) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150280
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.