Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hormone-sensitive Lipase/HSL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB11037253
Description
Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Hormone-sensitive Lipase/HSL | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| LIPE | |
| A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). [Swiss-Prot P15304] | |
| 0.05 mL | |
| Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 3991 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction