Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hormone-sensitive Lipase/HSL Antibody, Novus Biologicals™
SDP

Catalog No. p-5073692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11037253T 0.025 mL
NB11037253 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11037253T Supplier Novus Biologicals Supplier No. NB11037253SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Hormone-sensitive Lipase/HSL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive
Gene Symbols LIPE
Host Species Rabbit
Immunogen A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). [Swiss-Prot P15304]
Purification Method IgG purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3991
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.