Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hormone-sensitive Lipase/HSL, p Ser853 Antibody (SR2248), Novus Biologicals™
SDP

Catalog No. NB283097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB283097 100 μg
NB284293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB283097 Supplier Novus Biologicals Supplier No. NBP321545100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Hormone-sensitive Lipase/HSL Monoclonal antibody specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Hormone-sensitive Lipase/HSL
Applications Western Blot
Classification Monoclonal
Clone SR2248
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-Hormone-sensitive Lipase/HSL [Ser853] (Uniprot #: Q05469)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3991
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.