Learn More
Host Cell Factor 1/HCFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Host Cell Factor 1/HCFC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CFFC1 factor, HCF, HCF1HFC1, HCF-1VP16 accessory protein, host cell factor C1 (VP16-accessory protein), MGC70925, VCAFhost cell factor 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Host Cell Factor 1/HCFC1 (NP_005325.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.