Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences Hot start High Fidelity DNA Polymerase 2X Master Mix

Click to view available options
No. of Reactions:
100
500
Description
- G-biosciences hot start high-fidelity DNA Polymerase 2x master mix is ready to use premix which contains hot start high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers.
- The hot start high-fidelity DNA Polymerase possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
Specifications
Specifications
| Detection Method | qPCR, RT-PCR |
| GC-Rich PCR Performance | High |
| Reaction Speed | Fast or Standard |
| Product Type | DNA Polymerase Master Mix |
| For Use With (Application) | Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection |
| No. of Reactions | 100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction