Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Hot start High Fidelity DNA Polymerase 2X Master Mix
SCI_ED

Hot start High Fidelity DNA Polymerase 2X Master Mix is used for PCR, Cloning and Diagnostics purposes.

$172.00 - $645.00

Specifications

Detection Method qPCR, RT-PCR
GC-Rich PCR Performance High
Reaction Speed Fast or Standard
Product Type DNA Polymerase Master Mix
For Use With (Application) Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
S03144
SCI_ED
View Documents
G-Biosciences
7861973
500
Each for $645.00
Only null left
Add to Cart
 
S03145
SCI_ED
View Documents
G-Biosciences
7861972
100
Each for $172.00
Only null left
Add to Cart
 
Description

Description

  • G-biosciences hot start high-fidelity DNA Polymerase 2x master mix is ready to use premix which contains hot start high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers.
  • The hot start high-fidelity DNA Polymerase possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
Specifications

Specifications

qPCR, RT-PCR
Fast or Standard
Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
High
DNA Polymerase Master Mix
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.