Learn More
Description
- Primer sequestration effectively blocks greater than 99% of polymerase activity during setup effectively minimizing any nonspecific priming or primer-dimer formation
- Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
- Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
- The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
Specifications
Specifications
| Content And Storage | 1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant |
| Description | Preformulated and predispensed, freeze-dried PCR reagents mix including high-quality PuReTaq DNA Polymerase and a hot start activator protein, for increased specificity and reproducibility of PCR amplifications. |
| Format | Tube |
| Product Type | illustra Hot Start Mix RTG |
| Quantity | 100 Reactions |
| For Use With (Application) | Standard PCR |
| No. of Reactions | 100 |
| Volume | 500μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.